Learn More
AdipoGen Ferulenol (1 mg)
SDP
Supplier:  AdipoGen AGCN20011M001
Encompass_Preferred
Chemical. CAS: 6805-34-1. Formula: C24H30O3. MW: 366.5. Isolated from Ferula communis. Prenylated 4-hydroxycoumarin. Anti-tumor compound. Cytotoxic. Stimulator of tubulin polymerization in vitro. Inhibitor of colchicine binding to tubulin. Antitubercular antibiotic with potent antibacterial activity. Anti-coagulant, pro-haemorrhagic compound with higher activity than warfarin. Shows hepatocyte toxicity. Disrupts mitochondrial membrane potential.
Catalog No. 50-205-3358