Learn More
Abcam Ferulenol, ATP synthesis inhibitor, 10MG
Catalog No. 502705950
Encompass_Preferred
MW 366.5 Da, Purity >98%. ATP synthesis inhibitor. Potent antibiotic agent. Tubulin polymerization stimulator. Selective succinate ubiquinone reductase inhibitor. Induces mPTP opening and disrupts mitochondrial membrane potential. Shows cytotoxic effects against cancer cell lines. Shows more potent anticoagulant activity than warfarin (ab141979) in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5950 Supplier Abcam Supplier No. AB14425310MG
MW 366.5 Da, Purity >98%. ATP synthesis inhibitor. Potent antibiotic agent. Tubulin polymerization stimulator. Selective succinate ubiquinone reductase inhibitor. Induces mPTP opening and disrupts mitochondrial membrane potential. Shows cytotoxic effects against cancer cell lines. Shows more potent anticoagulant activity than warfarin (ab141979) in vivo. .
The product is subject to the following: Abcam Restricted Use Statement