Learn More
Abcam Ferulenol, ATP synthesis inhibitor, 5MG
Catalog No. 502705948
Encompass_Preferred
MW 366.5 Da, Purity >98%. ATP synthesis inhibitor. Potent antibiotic agent. Tubulin polymerization stimulator. Selective succinate ubiquinone reductase inhibitor. Induces mPTP opening and disrupts mitochondrial membrane potential. Shows cytotoxic effects against cancer cell lines. Shows more potent anticoagulant activity than warfarin (ab141979) in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5948 Supplier Abcam Supplier No. AB1442535MG
MW 366.5 Da, Purity >98%. ATP synthesis inhibitor. Potent antibiotic agent. Tubulin polymerization stimulator. Selective succinate ubiquinone reductase inhibitor. Induces mPTP opening and disrupts mitochondrial membrane potential. Shows cytotoxic effects against cancer cell lines. Shows more potent anticoagulant activity than warfarin (ab141979) in vivo. .
The product is subject to the following: Abcam Restricted Use Statement