Learn More
Medchemexpress LLC HY-N0060 100mg Medchemexpress, Ferulic acid CAS:1135-24-6 Purity:>98%
SDP
Catalog No. 501933374
Encompass_Preferred
Medchemexpress, HY-N0060 100mg Ferulic acid CAS:1135-24-6 Ferulic acid is a novel fibroblast growth factor receptor 1 ( FGFR1 ) inhibitor with IC 50 s of 3.78 and 12.5 μM for FGFR1 and FGFR2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3374 Supplier Medchemexpress LLC Supplier No. HYN0060100MG
Medchemexpress, HY-N0060 100mg Ferulic acid CAS:1135-24-6 Ferulic acid is a novel fibroblast growth factor receptor 1 ( FGFR1 ) inhibitor with IC 50 s of 3.78 and 12.5 μM for FGFR1 and FGFR2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.