Learn More
Cayman Chemical Ferulic Acid, Purity: ≥98%, CAS: 1135-24-6, 10 g
SDP
Catalog No. 502984951
Encompass_Preferred
A phenol with diverse biological activities; active against the yeast S. cerevisiae at 250 ppm and the bacteria E. coli, S. aureus, and B. cereus at 500 µg/ml; dietary administration reduces serum total cholesterol levels in a rat model of diet-induced hypercholesterolemia at 0.2% w/w; suppresses benzo(a)pyrene-induced forestomach tumor formation in mice
Catalog No. 50-298-4951 Supplier Cayman Chemical Supplier No. 1987110 G
A phenol with diverse biological activities; active against the yeast S. cerevisiae at 250 ppm and the bacteria E. coli, S. aureus, and B. cereus at 500 µg/ml; dietary administration reduces serum total cholesterol levels in a rat model of diet-induced hypercholesterolemia at 0.2% w/w; suppresses benzo(a)pyrene-induced forestomach tumor formation in mice