Learn More
Medchemexpress LLC Ferulic acid | 1135-24-6 | MFCD00004555 | 100.0 % | 194.18 | 25 MG
SDP
Catalog No. 5000430087
Encompass_Preferred
Ferulic acid (Standard) serves as the analytical standard for Ferulic acid, primarily intended for research and analytical applications. It is also recognized as a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor, exhibiting IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.
  • Analytical standard for Ferulic acid
  • Intended for research and analytical applications
  • Novel fibroblast growth factor receptor 1 (FGFR1) inhibitor
Catalog No. 50-004-30087 Supplier Medchemexpress LLC Supplier No. 25MGHYN0060R
Ferulic acid (Standard) serves as the analytical standard for Ferulic acid, primarily intended for research and analytical applications. It is also recognized as a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor, exhibiting IC50s of 3.78 and 12.5 μM for FGFR1 and FGFR2, respectively.
  • Analytical standard for Ferulic acid
  • Intended for research and analytical applications
  • Novel fibroblast growth factor receptor 1 (FGFR1) inhibitor