Learn More
Medchemexpress LLC Ferulic acid 4-O-sulfate | 86321-29-1 | 98.07% | 274.25 | 50 MG
SDP
Catalog No. 5000362953
Encompass_Preferred
Ferulic acid 4-O-sulfate is a metabolite of Ferulic acid that relaxes arteries and lowers blood pressure in mice. This compound causes significant concentration-dependent relaxations in mouse aorta, femoral and saphenous arteries.
  • Relaxes arteries and lowers blood pressure
  • Suitable for in vitro and in vivo studies
  • Significant concentration-dependent relaxations in mouse arteries
Catalog No. 50-003-62953 Supplier Medchemexpress LLC Supplier No. HY14732350MG
Ferulic acid 4-O-sulfate is a metabolite of Ferulic acid that relaxes arteries and lowers blood pressure in mice. This compound causes significant concentration-dependent relaxations in mouse aorta, femoral and saphenous arteries.
  • Relaxes arteries and lowers blood pressure
  • Suitable for in vitro and in vivo studies
  • Significant concentration-dependent relaxations in mouse arteries