Learn More
Medchemexpress LLC Ferulic acid 4-O-sulfate | 86321-29-1 | 98.1% | 274.25 | 1 ML
SDP
Catalog No. 5000359997
Encompass_Preferred
Ferulic acid 4-O-sulfate is a metabolite of ferulic acid. It relaxes arteries and lowers blood pressure in mice. This product is for research use only and not sold to patients.
  • Causes significant concentration-dependent relaxations in mouse aorta, femoral, and saphenous arteries (in vitro).
  • Relaxes arteries and lowers blood pressure in mice (in vivo).
  • Significantly decreases mean arterial pressure (MAP) immediately after intravenous injection in male Swiss mice.
Catalog No. 50-003-59997 Supplier Medchemexpress LLC Supplier No. HY14732310MM/1ML
Ferulic acid 4-O-sulfate is a metabolite of ferulic acid. It relaxes arteries and lowers blood pressure in mice. This product is for research use only and not sold to patients.
  • Causes significant concentration-dependent relaxations in mouse aorta, femoral, and saphenous arteries (in vitro).
  • Relaxes arteries and lowers blood pressure in mice (in vivo).
  • Significantly decreases mean arterial pressure (MAP) immediately after intravenous injection in male Swiss mice.