Learn More
Medchemexpress LLC Ferulic acid 4-O-sulfate | 86321-29-1 | 98.1% | 274.25 | 5 MG
SDP
Catalog No. 5000360380
Encompass_Preferred
Ferulic acid 4-O-sulfate is a metabolite of Ferulic acid. It has been shown to relax arteries and lower blood pressure in mice. This compound causes significant concentration-dependent relaxations in mouse aorta, femoral, and saphenous arteries at concentrations of 0.1-30 μM.
  • Metabolite of Ferulic acid
  • Relaxes arteries
  • Lowers blood pressure in mice
  • Causes concentration-dependent relaxations in mouse aorta, femoral, and saphenous arteries
Catalog No. 50-003-60380 Supplier Medchemexpress LLC Supplier No. HY1473235MG
Ferulic acid 4-O-sulfate is a metabolite of Ferulic acid. It has been shown to relax arteries and lower blood pressure in mice. This compound causes significant concentration-dependent relaxations in mouse aorta, femoral, and saphenous arteries at concentrations of 0.1-30 μM.
  • Metabolite of Ferulic acid
  • Relaxes arteries
  • Lowers blood pressure in mice
  • Causes concentration-dependent relaxations in mouse aorta, femoral, and saphenous arteries