Learn More
Selleck Chemical LLC Ferulic Acid S2300-10mM/1mL
SDP
Catalog No. 503111779
Encompass_Preferred
Ferulic Acid (Fumalic) is a hydroxycinnamic acid and a type of organic compound found in the Ferula assafoetida L or Ligusticum chuanxiong Ferulic acid is a novel fibroblast growth factor receptor (FGFR) inhibitor with IC50s of 3 78 M and 12 5 M for FGFR1 and FGFR2 respectively
Catalog No. 50-311-1779 Supplier Selleck Chemical LLC Supplier No. S230010MM/1ML
Ferulic Acid (Fumalic) is a hydroxycinnamic acid and a type of organic compound found in the Ferula assafoetida L or Ligusticum chuanxiong Ferulic acid is a novel fibroblast growth factor receptor (FGFR) inhibitor with IC50s of 3 78 M and 12 5 M for FGFR1 and FGFR2 respectively