Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferulic acid acyl-β-D-glucoside | 7196-71-6 | 1 ML
SDP

Catalog No. 5000417304
Encompass_Preferred

Ferulic acid acyl-β-D-glucoside is a metabolite of Ferulic Acid that can be isolated from flaxseed extract. It is identified as a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor and is intended for research use only.

  • Metabolite of Ferulic Acid
  • Can be isolated from flaxseed extract
  • Inhibits fibroblast growth factor receptor 1 (FGFR1) and FGFR2
  • Purity: 99.33%
  • Appearance: White to off-white solid
  • Soluble in water (50 mg/mL)
  • Soluble in PBS (25 mg/mL)
  • Initial source from plants (Rosaceae, Aruncus sylvester Kostel.)

Catalog No. 50-004-17304 Supplier Medchemexpress LLC Supplier No. HYN771510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferulic acid acyl-β-D-glucoside is a metabolite of Ferulic Acid that can be isolated from flaxseed extract. It is identified as a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor and is intended for research use only.

  • Metabolite of Ferulic Acid
  • Can be isolated from flaxseed extract
  • Inhibits fibroblast growth factor receptor 1 (FGFR1) and FGFR2
  • Purity: 99.33%
  • Appearance: White to off-white solid
  • Soluble in water (50 mg/mL)
  • Soluble in PBS (25 mg/mL)
  • Initial source from plants (Rosaceae, Aruncus sylvester Kostel.)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.