Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferulic acid acyl-β-D-glucoside | 7196-71-6 | 25 MG
SDP

Catalog No. 5000421838
Encompass_Preferred

Ferulic acid acyl-β-D-glucoside is a metabolite of Ferulic Acid, isolated from flaxseed extract. It acts as a fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50 values of 3.78 μM for FGFR1 and 12.5 μM for FGFR2.

  • Metabolite of Ferulic Acid
  • Can be isolated from flaxseed extract
  • Functions as a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor
  • Purity of 99.33%
  • Molecular weight: 356.32
  • Molecular formula: C16H20O9
  • Appearance: White to off-white solid

Catalog No. 50-004-21838 Supplier Medchemexpress LLC Supplier No. 25MGHYN7715
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferulic acid acyl-β-D-glucoside is a metabolite of Ferulic Acid, isolated from flaxseed extract. It acts as a fibroblast growth factor receptor 1 (FGFR1) inhibitor with IC50 values of 3.78 μM for FGFR1 and 12.5 μM for FGFR2.

  • Metabolite of Ferulic Acid
  • Can be isolated from flaxseed extract
  • Functions as a novel fibroblast growth factor receptor 1 (FGFR1) inhibitor
  • Purity of 99.33%
  • Molecular weight: 356.32
  • Molecular formula: C16H20O9
  • Appearance: White to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.