Learn More
Cayman Chemical Ferulic Acid ethyl ester, Purity: ≥98%, CAS: 4046-02-0, 10 g
SDP
Catalog No. 502951988
Encompass_Preferred
A lipophilic derivative of ferulic acid, with increased ability to cross cell membranes; less DPPH radical scavenging capacity than ferulic acid (IC50 = 66.7 μM for FAEE vs. 44.6 μM for ferulic acid); increases the expression of HO-1, Hsp70, and Hsp72, providing neuroprotection against amyloid β-peptide
Catalog No. 50-295-1988 Supplier Cayman Chemical Supplier No. 1188010 G
A lipophilic derivative of ferulic acid, with increased ability to cross cell membranes; less DPPH radical scavenging capacity than ferulic acid (IC50 = 66.7 μM for FAEE vs. 44.6 μM for ferulic acid); increases the expression of HO-1, Hsp70, and Hsp72, providing neuroprotection against amyloid β-peptide