Learn More
Cayman Chemical Ferulic Acid methyl ester, Purity: ≥98%, CAS: 2309-07-1, 1 g
SDP
Catalog No. 502966832
Encompass_Preferred
A lipophilic derivative of ferulic acid, demonstrating increased ability to cross cell membranes; at 10-25 µg/ml, inhibits the release of pro-inflammatory cytokines, blocks the expression of COX-2, and reduces NO generation from LPS-stimulated macrophages
Catalog No. 50-296-6832 Supplier Cayman Chemical Supplier No. 178811 G
A lipophilic derivative of ferulic acid, demonstrating increased ability to cross cell membranes; at 10-25 µg/ml, inhibits the release of pro-inflammatory cytokines, blocks the expression of COX-2, and reduces NO generation from LPS-stimulated macrophages