Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferulic acid methyl ester | 2309-07-1 | 99.9% | 208.21 g/mol | C11H12O4 | 1 G
SDP

Catalog No. 5000300850
Encompass_Preferred

Ferulic acid methyl ester (methyl ferulate) is the methyl ester derivative of ferulic acid used as a research reagent. It is reported as a p38 MAPK inhibitor with anti-inflammatory and antioxidant properties and is applied in studies of autophagy and neurological disease. The compound is cell membrane and brain permeable and modulates COX-2, p-p38, and p-JNK signaling.

  • CAS number 2309-07-1
  • Chemical formula C11H12O4
  • Molecular weight 208.21 g/mol
  • Typical purity ~99.9% (rounded)
  • Available as solid and 10 mM solution in DMSO
  • Reported p38 MAPK inhibitor with anti-inflammatory activity
  • Useful for autophagy and neurological disease research

Catalog No. 50-003-00850 Supplier Medchemexpress LLC Supplier No. HYW0186431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferulic acid methyl ester (methyl ferulate) is the methyl ester derivative of ferulic acid used as a research reagent. It is reported as a p38 MAPK inhibitor with anti-inflammatory and antioxidant properties and is applied in studies of autophagy and neurological disease. The compound is cell membrane and brain permeable and modulates COX-2, p-p38, and p-JNK signaling.

  • CAS number 2309-07-1
  • Chemical formula C11H12O4
  • Molecular weight 208.21 g/mol
  • Typical purity ~99.9% (rounded)
  • Available as solid and 10 mM solution in DMSO
  • Reported p38 MAPK inhibitor with anti-inflammatory activity
  • Useful for autophagy and neurological disease research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.