Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferulic acid methyl ester | 2309-07-1 | MFCD00017208 | 99.9% | 208.21 g/mol | C11H12O4 | 1 ML
SDP

Catalog No. 5000302025
Encompass_Preferred

Ferulic acid methyl ester is a methyl ester derivative of ferulic acid used as a research reagent in biochemical and cell-based studies of MAPK signaling, oxidative stress, and autophagy. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) to support consistent dosing in in vitro experiments.

  • Ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO and compatible with common in vitro vehicles.
  • Stability data provided for powder and solution storage.
  • Supplied with datasheet and SDS for experimental support.

Catalog No. 50-003-02025 Supplier Medchemexpress LLC Supplier No. HYW01864310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferulic acid methyl ester is a methyl ester derivative of ferulic acid used as a research reagent in biochemical and cell-based studies of MAPK signaling, oxidative stress, and autophagy. It is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) to support consistent dosing in in vitro experiments.

  • Ready-to-use 10 mM solution in DMSO for convenient dosing.
  • High purity suitable for biochemical and cellular assays.
  • Soluble in DMSO and compatible with common in vitro vehicles.
  • Stability data provided for powder and solution storage.
  • Supplied with datasheet and SDS for experimental support.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.