Learn More
Medchemexpress LLC Ferulic acid methyl ester | 2309-07-1 | MFCD00017208 | 99.9% | 208.21 g/mol | C11H12O4 | 10 G
SDP
Catalog No. 5000300852
Encompass_Preferred
Ferulic acid methyl ester is the methyl ester derivative of ferulic acid used as a research reagent. It shows antioxidant and anti-inflammatory properties, is reported to inhibit p38 MAPK and modulate autophagy, and is suitable for biochemical and cell-based studies; batch-specific purity and safety documentation are available from the manufacturer.
  • Methyl ester derivative of ferulic acid with antioxidant activity.
  • Reported inhibitor of p38 MAPK and modulator of autophagy pathways.
  • Cell membrane and brain permeable, suitable for cell-based assays.
  • High batch-specific purity with certificate of analysis and SDS documentation available.
  • Available in multiple pack sizes for research use, including a 10 g pack.
Catalog No. 50-003-00852 Supplier Medchemexpress LLC Supplier No. HYW01864310G
Ferulic acid methyl ester is the methyl ester derivative of ferulic acid used as a research reagent. It shows antioxidant and anti-inflammatory properties, is reported to inhibit p38 MAPK and modulate autophagy, and is suitable for biochemical and cell-based studies; batch-specific purity and safety documentation are available from the manufacturer.
  • Methyl ester derivative of ferulic acid with antioxidant activity.
  • Reported inhibitor of p38 MAPK and modulator of autophagy pathways.
  • Cell membrane and brain permeable, suitable for cell-based assays.
  • High batch-specific purity with certificate of analysis and SDS documentation available.
  • Available in multiple pack sizes for research use, including a 10 g pack.