Learn More
Medchemexpress LLC Ferulic acid methyl ester | 2309-07-1 | MFCD00017208 | 99.9% | 208.21 g/mol | C11H12O4 | 25 G
SDP
Catalog No. 5000303870
Encompass_Preferred
Ferulic acid methyl ester is the methyl ester derivative of ferulic acid used in biochemical and pharmacological research. It functions as a lipophilic antioxidant with reported activity in pathways such as p38 MAPK and autophagy, and is commonly used in in vitro and cell-based studies.
  • High purity suitable for research applications.
  • Lipophilic antioxidant and cell-permeable compound.
  • Solid, off-white to pink appearance for easy handling.
  • Available in multiple pack sizes, including 25 g.
  • Stable when stored according to supplier recommendations (powder: -20°C or 4°C).
Catalog No. 50-003-03870 Supplier Medchemexpress LLC Supplier No. 25GHYW018643
Ferulic acid methyl ester is the methyl ester derivative of ferulic acid used in biochemical and pharmacological research. It functions as a lipophilic antioxidant with reported activity in pathways such as p38 MAPK and autophagy, and is commonly used in in vitro and cell-based studies.
  • High purity suitable for research applications.
  • Lipophilic antioxidant and cell-permeable compound.
  • Solid, off-white to pink appearance for easy handling.
  • Available in multiple pack sizes, including 25 g.
  • Stable when stored according to supplier recommendations (powder: -20°C or 4°C).