Learn More
Selleck Chemical LLC Ferulic acid methyl ester S5451-25mg
SDP
Catalog No. 503110280
Encompass_Preferred
Ferulic acid methyl ester (Methyl Ferulate Methyl 4 -hydroxy-3 -methoxycinnamate) is a lipophilic derivative of ferulic acid which is a hydroxycinnamic acid that is abundant in plants It shows the strongest antioxidant activity and can protect against inflammation and cancer
Catalog No. 50-311-0280 Supplier Selleck Chemical LLC Supplier No. S545125MG
Ferulic acid methyl ester (Methyl Ferulate Methyl 4 -hydroxy-3 -methoxycinnamate) is a lipophilic derivative of ferulic acid which is a hydroxycinnamic acid that is abundant in plants It shows the strongest antioxidant activity and can protect against inflammation and cancer