Learn More
Abcam Ferulic acid methyl ester, Antio x idant agent, 25G
Catalog No. 502706100
Encompass_Preferred
MW 208.21 Da, Purity >99%. Antioxidant agent. Ferulic acid (ab141932) derivative. Putative chemotherapeutic agent. Shows antithrombotic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6100 Supplier Abcam Supplier No. AB14362025G
MW 208.21 Da, Purity >99%. Antioxidant agent. Ferulic acid (ab141932) derivative. Putative chemotherapeutic agent. Shows antithrombotic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement