Learn More
Medchemexpress LLC Ferulic acid methyl ester (methyl ferulate) | 2309-07-1 | MFCD00017208 | 100.0% | 208.21 g·mol⁻¹ | C11H12O4 | 5 G
SDP
Catalog No. 5000301882
Encompass_Preferred
Ferulic acid methyl ester is the methyl ester derivative of ferulic acid supplied for research use. It is reported to be cell membrane- and brain-permeable, with antioxidant and anti-inflammatory activity, and has been used to study MAPK signaling and COX-2 expression. The compound is provided at high purity for biochemical and cellular research applications.
  • Methyl ester derivative of ferulic acid
  • Antioxidant and anti-inflammatory activity
  • Cell membrane and brain permeable
  • Modulates MAPK signaling and COX-2 expression
  • High purity suitable for research applications
Catalog No. 50-003-01882 Supplier Medchemexpress LLC Supplier No. HYW0186435G
Ferulic acid methyl ester is the methyl ester derivative of ferulic acid supplied for research use. It is reported to be cell membrane- and brain-permeable, with antioxidant and anti-inflammatory activity, and has been used to study MAPK signaling and COX-2 expression. The compound is provided at high purity for biochemical and cellular research applications.
  • Methyl ester derivative of ferulic acid
  • Antioxidant and anti-inflammatory activity
  • Cell membrane and brain permeable
  • Modulates MAPK signaling and COX-2 expression
  • High purity suitable for research applications