Learn More
Cayman Chemical Ferulic Acid methyl ester, Purity: ≥98%, CAS: 2309-07-1, 500 mg
SDP
Catalog No. 502967394
Encompass_Preferred
A lipophilic derivative of ferulic acid, demonstrating increased ability to cross cell membranes; at 10-25 µg/ml, inhibits the release of pro-inflammatory cytokines, blocks the expression of COX-2, and reduces NO generation from LPS-stimulated macrophages
Catalog No. 50-296-7394 Supplier Cayman Chemical Supplier No. 17881500 MG
A lipophilic derivative of ferulic acid, demonstrating increased ability to cross cell membranes; at 10-25 µg/ml, inhibits the release of pro-inflammatory cytokines, blocks the expression of COX-2, and reduces NO generation from LPS-stimulated macrophages