Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ferulic acid (standard) | 1135-24-6 | 100.0% | 10 MG
SDP

Catalog No. 5000297442
Encompass_Preferred

Ferulic acid (standard) is an analytical reference standard intended for qualitative and quantitative research, method development, and instrument calibration in chromatography and mass spectrometry. It is provided at defined high purity for reproducible analytical results.

  • Analytical standard for HPLC, GC, and MS applications.
  • High purity, suitable for quantitative analysis.
  • CAS number 1135-24-6 for unambiguous identification.
  • Molecular weight 194.18 and formula C10H10O4.
  • Available in small quantities suitable for laboratory method development and calibration.

Catalog No. 50-002-97442 Supplier Medchemexpress LLC Supplier No. HYN0060R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ferulic acid (standard) is an analytical reference standard intended for qualitative and quantitative research, method development, and instrument calibration in chromatography and mass spectrometry. It is provided at defined high purity for reproducible analytical results.

  • Analytical standard for HPLC, GC, and MS applications.
  • High purity, suitable for quantitative analysis.
  • CAS number 1135-24-6 for unambiguous identification.
  • Molecular weight 194.18 and formula C10H10O4.
  • Available in small quantities suitable for laboratory method development and calibration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.