Learn More
AdipoGen Ferutinin (high purity) (5 mg)
SDP
Supplier:  AdipoGen AGCN20007M005
Encompass_Preferred
Chemical. CAS: 41743-44-6. Formula: C22H30O4. MW: 358.5. Semisynthetic. Potent, naturally occuring non-steroid estrogenic compound. Tool to promote mitochondrial calcium overload and to promote calcium-dependent opening of the mitochondrial permeability transition pore (mPTP). Strong agonist for estrogen receptor (ER)alpha and agonist/antagonist for ERbeta. Calcium ionophoretic Apoptosis modulator. Antiproliferative. Increases nitric oxide synthase activity and phosphoinositides breakdown in nervous tissue. Shows aphrodisiac and anti-sexual impotence activities. Anti-osteoporotic. Prevents osteoporosis caused by severe estrogen deficiency. Modest AChE inhibitor.
Catalog No. 50-205-3353