Learn More
Aviva Systems Biology Corp FES Antibody - N-terminal region (ARP54607_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54607_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FES. It was validated on Western Blot. . Alternate names: FPSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-946