Learn More
Eurofins Discoverx Fes/Fps, active, 250 µg. (14-473M)
Supplier:  Eurofins Discoverx 14473M
Encompass
N-terminal 6xHis-tagged recombinant, human full length Fes. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8840