Learn More
Medchemexpress LLC Fesomersen (sodium) | 2380288-85-5 | 98.7% | 9113.01 | 1 MG
SDP
Catalog No. 5000351220
Encompass_Preferred
Fesomersen (sodium) is an antisense oligonucleotide designed to inhibit the production of Factor XI. It is intended for research use only and is not sold to patients.
  • Inhibits Factor XI production
  • Suitable for laboratory chemical applications
  • For research use only
Catalog No. 50-003-51220 Supplier Medchemexpress LLC Supplier No. HY139788A1MG
Fesomersen (sodium) is an antisense oligonucleotide designed to inhibit the production of Factor XI. It is intended for research use only and is not sold to patients.
  • Inhibits Factor XI production
  • Suitable for laboratory chemical applications
  • For research use only