Learn More
Apexbio Technology LLC Fesoterodine Fumarate 286930-03-8 5mg
SDP
Catalog No. 5000092708
Encompass_Preferred
Fesoterodine Fumarate (CAS 286930-03-8) is a synthetic antimuscarinic agent that acts as a competitive antagonist at muscarinic acetylcholine receptors particularly the M3 subtype in the bladder detrusor muscle By inhibiting these receptors the compound modulates involuntary bladder contractions reducing symptoms associated with overactive bladder syndrome such as urgency and frequency Fesoterodine Fumarate is utilized in biomedical research to study cholinergic signaling mechanisms involved in urinary function and to explore pharmacological interventions targeting muscarinic pathways in urological disorders
Catalog No. 50-000-92708 Supplier Apexbio Technology LLC Supplier No. B16185
Fesoterodine Fumarate (CAS 286930-03-8) is a synthetic antimuscarinic agent that acts as a competitive antagonist at muscarinic acetylcholine receptors particularly the M3 subtype in the bladder detrusor muscle By inhibiting these receptors the compound modulates involuntary bladder contractions reducing symptoms associated with overactive bladder syndrome such as urgency and frequency Fesoterodine Fumarate is utilized in biomedical research to study cholinergic signaling mechanisms involved in urinary function and to explore pharmacological interventions targeting muscarinic pathways in urological disorders