Learn More
Medchemexpress LLC Fesoterodine fumarate | 286930-03-8 | 99.5% | C30H41NO7 | 50 MG
SDP
Catalog No. 5000432993
Encompass_Preferred
Fesoterodine fumarate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist. It is primarily used for the treatment of overactive bladder (OAB), where it helps decrease micturition frequency, urgency severity, and urgency incontinence episodes, while increasing the volume voided.
  • Orally active, nonsubtype selective mAChR antagonist
  • Treats overactive bladder (OAB)
  • Decreases micturition frequency, urgency severity, and urgency incontinence episodes
  • Increases volume voided with each micturition
  • Reduces micturition pressure
  • Increases bladder capacity and intercontraction intervals (ICIs)
Catalog No. 50-004-32993 Supplier Medchemexpress LLC Supplier No. HYA003050MG
Fesoterodine fumarate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist. It is primarily used for the treatment of overactive bladder (OAB), where it helps decrease micturition frequency, urgency severity, and urgency incontinence episodes, while increasing the volume voided.
  • Orally active, nonsubtype selective mAChR antagonist
  • Treats overactive bladder (OAB)
  • Decreases micturition frequency, urgency severity, and urgency incontinence episodes
  • Increases volume voided with each micturition
  • Reduces micturition pressure
  • Increases bladder capacity and intercontraction intervals (ICIs)