Learn More
Medchemexpress LLC Fesoterodine (fumarate) | 286930-03-8 | MFCD09835695 | ≥98% | 574.67 | 10 MG
SDP
Catalog No. 5000365136
Encompass_Preferred
Fesoterodine fumarate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist. It is used for the overactive bladder (OAB).
  • Orally active
  • Nonsubtype selective
  • Competitive muscarinic receptor (mAChR) antagonist
  • Targets mAChR
  • Used for overactive bladder (OAB)
  • Research areas include neurological disease, metabolic disease, and cancer
Catalog No. 50-003-65136 Supplier Medchemexpress LLC Supplier No. 10MGHYA0030
Fesoterodine fumarate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist. It is used for the overactive bladder (OAB).
  • Orally active
  • Nonsubtype selective
  • Competitive muscarinic receptor (mAChR) antagonist
  • Targets mAChR
  • Used for overactive bladder (OAB)
  • Research areas include neurological disease, metabolic disease, and cancer