Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fesoterodine L-mandelate | 1206695-46-6 | 98.9% | C34H45NO6 | 10 MM * 1 ML
SDP

Catalog No. 5000434614
Encompass_Preferred

Fesoterodine L-mandelate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist used for the treatment of overactive bladder (OAB).

  • Orally active
  • Nonsubtype selective
  • Competitive muscarinic receptor (mAChR) antagonist
  • Used for the treatment of overactive bladder (OAB)
  • Exhibits pKi values of 8.0, 7.7, 7.4, 7.3, and 7.5 for M1, M2, M3, M4, and M5 receptors, respectively.

Catalog No. 50-004-34614 Supplier Medchemexpress LLC Supplier No. HY70053A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fesoterodine L-mandelate is an orally active, nonsubtype selective, competitive muscarinic receptor (mAChR) antagonist used for the treatment of overactive bladder (OAB).

  • Orally active
  • Nonsubtype selective
  • Competitive muscarinic receptor (mAChR) antagonist
  • Used for the treatment of overactive bladder (OAB)
  • Exhibits pKi values of 8.0, 7.7, 7.4, 7.3, and 7.5 for M1, M2, M3, M4, and M5 receptors, respectively.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.