Learn More
Med Vet International FESSA Tubular Fixator System, 6mm Allen Wrench, J1050D
Catalog No. 5000583411
Encompass_Preferred
This system is widely used in Europe and offers several advantages over conventional fixator systems. The connecting tubes or rods use a simple hexhead set screw as the pin holding or clamping mechanism. The system is very strong for its light weight. The smaller tubes will accept the smallest of K-wire making it extremely useful in fracture management of small exotics and cats.
Catalog No. 50-005-83411 Supplier Med Vet International Supplier No. JOR1050D
This system is widely used in Europe and offers several advantages over conventional fixator systems. The connecting tubes or rods use a simple hexhead set screw as the pin holding or clamping mechanism. The system is very strong for its light weight. The smaller tubes will accept the smallest of K-wire making it extremely useful in fracture management of small exotics and cats.