Learn More
Med Vet International Fessa Tubular Fixator System, 8mm Locking Screws, J1051E
Catalog No. 5000583602
Encompass_Preferred
This system is widely used in Europe and offers several advantages over conventional fixator systems. The connecting tubes or rods use a simple hexhead set screw as the pin holding or clamping mechanism. The system is very strong for its light weight. The smaller tubes will accept the smallest of K-wire making it extremely useful in fracture management of small exotics and cats.
Catalog No. 50-005-83602 Supplier Med Vet International Supplier No. JOR1051E
This system is widely used in Europe and offers several advantages over conventional fixator systems. The connecting tubes or rods use a simple hexhead set screw as the pin holding or clamping mechanism. The system is very strong for its light weight. The smaller tubes will accept the smallest of K-wire making it extremely useful in fracture management of small exotics and cats.