Learn More
BIORELEVANT COM LTD BIORELEVANT COM LTD Mini Prep Kit, FeSSIF, 6 x 100 mL, pH 5.0
Catalog No. NC3993692
Encompass
This mini prep kit allows for the convenient preparation of small volumes of simulated intestinal fluid. It accurately mimics the fed prandial state found in the small intestine, providing a consistent pH of 5.0 for reliable experimental conditions.
  • New product offering for simplified preparation.
  • Designed for preparing small volumes of simulated intestinal fluid.
  • Simulates the fed prandial state in small intestinal fluid.
  • Maintains a pH of 5.0.
  • Supplied in a 6 x 100 mL format.
Catalog No. NC3993692 Supplier BIORELEVANT COM LTD Supplier No. FESSIFMP
This mini prep kit allows for the convenient preparation of small volumes of simulated intestinal fluid. It accurately mimics the fed prandial state found in the small intestine, providing a consistent pH of 5.0 for reliable experimental conditions.
  • New product offering for simplified preparation.
  • Designed for preparing small volumes of simulated intestinal fluid.
  • Simulates the fed prandial state in small intestinal fluid.
  • Maintains a pH of 5.0.
  • Supplied in a 6 x 100 mL format.