Learn More
Kalen Biomedical LLC Fetal Bovine Lipoprotein Depleted Serum, 50 mL, < 1.25 g/mL density, 35.0 mg/mL protein concentration, ultracentrifuged, aseptically packaged
Catalog No. NC9764296
Encompass_Preferred
Kalen Biomedical LLC's Lipoprotein Depleted Fetal Bovine Serum (LPDS) is a specialized cell culture supplement tailored for lipid homeostasis research. This serum is produced through ultracentrifugation, resulting in a significant reduction of lipoproteins to a density of less than 1.25 g/mL, and is filtered to ensure purity. It is aseptically packaged and stored frozen, making it ideal for cellular applications that require lipid-deficient conditions. Key features include:
  • Depletes lipoproteins effectively
  • Protein concentration of approximately 35.0 mg/mL
  • Volume of 50 mL
  • Aseptically packaged to ensure sterility
  • Ultra-filtered for enhanced purity
  • Geographically available in North America only
  • Shipping scheduled for Wednesdays
  • Stored at -20°C
  • 40-fold cholesterol reduction compared to standard fetal bovine serum
  • Iron content comparable to Sigma FBS
Catalog No. NC9764296 Supplier Kalen Biomedical LLC Supplier No. 8801005
Kalen Biomedical LLC's Lipoprotein Depleted Fetal Bovine Serum (LPDS) is a specialized cell culture supplement tailored for lipid homeostasis research. This serum is produced through ultracentrifugation, resulting in a significant reduction of lipoproteins to a density of less than 1.25 g/mL, and is filtered to ensure purity. It is aseptically packaged and stored frozen, making it ideal for cellular applications that require lipid-deficient conditions. Key features include:
  • Depletes lipoproteins effectively
  • Protein concentration of approximately 35.0 mg/mL
  • Volume of 50 mL
  • Aseptically packaged to ensure sterility
  • Ultra-filtered for enhanced purity
  • Geographically available in North America only
  • Shipping scheduled for Wednesdays
  • Stored at -20°C
  • 40-fold cholesterol reduction compared to standard fetal bovine serum
  • Iron content comparable to Sigma FBS