Learn More
Atlas Biologicals EqualFetal Bovine Serum, HEAT INACTIVATED, US Origin, 500mL
SDP
Catalog No. NC1808866
Encompass
Derived from bovine animals after birth that have been tested to meet FBS specifications. EqualFetal is highly similar to FBS, intended to be used interchangeably, and expected to perform the same on select cell lines. In fact, USP and ISIA FBS specifications are equivalent to EqualFetal. EqualFetal is comprised of PURE bovine serum and contains no additives.
Catalog No. NC1808866 Supplier Atlas Biologicals Supplier No. EF0500A (HI)
Derived from bovine animals after birth that have been tested to meet FBS specifications. EqualFetal is highly similar to FBS, intended to be used interchangeably, and expected to perform the same on select cell lines. In fact, USP and ISIA FBS specifications are equivalent to EqualFetal. EqualFetal is comprised of PURE bovine serum and contains no additives.