Learn More
ATCC Fetal Bovine Serum (100 mL)
Catalog No. 502384615
Encompass
Fetal Bovine Serum (FBS) is triple filtered through 0.1 μm filters. Each lot of fetal bovine serum is tested for sterility and for the ability to support the growth of several different cell lines using both sequential growth curves and plating efficiencies. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA inspected-abattoirs located in the United States. This product is 100 mL. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-4615 Supplier ATCC Supplier No. 302021
Fetal Bovine Serum (FBS) is triple filtered through 0.1 μm filters. Each lot of fetal bovine serum is tested for sterility and for the ability to support the growth of several different cell lines using both sequential growth curves and plating efficiencies. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA inspected-abattoirs located in the United States. This product is 100 mL. It has applications for cell culture, cell growth, and viability.