Learn More
ATCC Fetal Bovine Serum, 500mL, Frozen
Supplier:  ATCC 302020/DEL
Encompass
It is triple filtered through 0.1-um filters. Each lot of fetal bovine serum is tested for sterility and for the ability to support the growth of several different cell lines using both sequential growth curves and plating efficiencies. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA-inspected abattoirs located in the United States. For use by FOR-PROFIT companies only; NON-PROFIT companies use 50189023NP. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-189-023FP