Learn More
Atlas Biologicals Fetal Plus Bovine Serum, US Origin, 500mL
SDP
Catalog No. NC0174356
Encompass
Fetal Plus is a proprietary combination of US origin FBS and EquaFETAL. Fetal Plus is derived from 100% U.S. Origin bovine serum Fetal Plus supports a wide variety of cell types. The formulation is proprietary and the exclusive property of Atlas Biologicals. Produced from raw materials collected from approved facilities within the contiguous United States that has undergone inspection by the United States Department of Agriculture. Manufactured utilizing current Good Manufacturing Practices (cGMP). Triple 0.1 micron filtered.
Catalog No. NC0174356 Supplier Atlas Biologicals Supplier No. FP0500A
Fetal Plus is a proprietary combination of US origin FBS and EquaFETAL. Fetal Plus is derived from 100% U.S. Origin bovine serum Fetal Plus supports a wide variety of cell types. The formulation is proprietary and the exclusive property of Atlas Biologicals. Produced from raw materials collected from approved facilities within the contiguous United States that has undergone inspection by the United States Department of Agriculture. Manufactured utilizing current Good Manufacturing Practices (cGMP). Triple 0.1 micron filtered.