Learn More
ATCC Fetal Bovine Serum (500 mL)
Catalog No. 502383771
Encompass
Fetal Bovine Serum (FBS) is triple filtered through 0.1 ?m filters. Each lot of fetal bovine serum is tested for sterility and for the ability to support the growth of several different cell lines using both sequential growth curves and plating efficiencies. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA inspected-abattoirs located in the United States. This product is 500 mL. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3771 Supplier ATCC Supplier No. 302020
 Restricted Item
Add to Cart
Add to Cart
Fetal Bovine Serum (FBS) is triple filtered through 0.1 ?m filters. Each lot of fetal bovine serum is tested for sterility and for the ability to support the growth of several different cell lines using both sequential growth curves and plating efficiencies. Fetal bovine serum is manufactured from fetal bovine blood collected in USDA inspected-abattoirs located in the United States. This product is 500 mL. It has applications for cell culture, cell growth, and viability.