Learn More
Sigma Aldrich Fine Chemicals Biosciences fetal Bovine Serum Canada
Catalog No. 502901585
Encompass
fetal bovine serum is the main component present in almost all media used for the culture of animal cells. It provides serum additives to the cell culture medium which is essential for supporting growth of a variety of cell types. It is rich in growth factors and protein but low in³-globulin content.
Catalog No. 50-290-1585 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F1051100ML
fetal bovine serum is the main component present in almost all media used for the culture of animal cells. It provides serum additives to the cell culture medium which is essential for supporting growth of a variety of cell types. It is rich in growth factors and protein but low in³-globulin content.