Learn More
Novus Biologicals™ Fetal Bovine Serum - Charcoal/Dextran Treated, Heat Inactivated

Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Description | Fetal Bovine Serum - Charcoal/Dextran Treated, Heat Inactivated |
| Species | Bovine |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| For Use With (Application) | Serum |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.