Learn More
Alkali Scientific ASI™ Fetal Bovine Serum (FBS), Tetracycline Approved, 500mL
SDP
Catalog No. 503028908
Encompass_Preferred
CellPro™ Tetracycline is an antibiotic commonly found in Fetal Bovine Serum which can cause problems for researchers using sensitive tet-inducing expression systems. Processed sera is screened for the presence of Tetracycline. This FBS has no detectable level of Tetracycline present. Virus-screened Mycoplasma-tested Triple 0.1 Micron Filtered Low Hemoglobin lots (<15mg/dL) may be available. Low Endotoxin lots (<1.0EU/mL) may be available. This product ships on dry ice.
Catalog No. 50-302-8908 Supplier Alkali Scientific Supplier No. FB76
CellPro™ Tetracycline is an antibiotic commonly found in Fetal Bovine Serum which can cause problems for researchers using sensitive tet-inducing expression systems. Processed sera is screened for the presence of Tetracycline. This FBS has no detectable level of Tetracycline present. Virus-screened Mycoplasma-tested Triple 0.1 Micron Filtered Low Hemoglobin lots (<15mg/dL) may be available. Low Endotoxin lots (<1.0EU/mL) may be available. This product ships on dry ice.