Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Fetal Bovine Serum (FBS)

Catalog No. 092910154 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-291-0154 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-291-0154 Supplier MP Biomedicals Supplier No. 092910154
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in a broad range of cell culture applications for promoting/enhancing growth/survival.

  • FBS provides many non-defined growth promoting and survival enhancing factors to cells in culture.
  • For cell culture, mammalian cell culture, source of growth promoting and survival enhancing factor.
  • Golden brown liquid.
  • Sterile.
TRUSTED_SUSTAINABILITY

Specifications

Color Golden brown
Content And Storage Store at -20°C
Form Liquid
Quantity 500 mL
Concentration Protein: 3.4 - 4.6%
Sterility Sterile
Formulation Liquid; Glucose: 137 mg/dL; Sodium: 140 meq/L; Potassium: 10.6 meq/L; Chloride: 103 meq/L; B.U.N.: 15 mg/dL; Creatinine: 2.7 mg/dL; Uric Acid: 2.3 mg/dL; Calcium: 14.0 mg/dL; Phosphorus: 9.9 mg/dL; Albumin: 1.9 gm/dL; Total Bilirubin: 0.2 mg/dL; ALT (SGPT): 5 IU/L; Alkaline phosphatase: 261 IU/L; AST (SGOT): 43 IU/L; LDH: 556 IU/L; Cholesterol: 31 mg/dL; Triglycerides: 57 mg/dL.
For Use With (Application) Cell culture. Mammalian cell culture. Source of growth promoting and survival enhancing factor.
pH pH 6.7-7.6
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.