Learn More
Sigma Aldrich Fine Chemicals Biosciences fetal Bovine Serum fetal B
Catalog No. 502904610
Encompass
Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro. fetal Bovine Serum (fBS) is the most common serum used to supplement cell culture media due to its high nutritional content. Although it is relatively low in protein fBS is effective in promoting and sustaining growth of vertebrate mammalian and insect cells. IgG is the most common immunoglobulin present in fBS. Elevated levels of IgG may cause problems within certain recombinant protein or monoclonal antibody purification processes. for applications where IgG may interfere or cause issues we have depleted IgG levels in this fetal bovine serum by sequential chromatography down to a level of 5 ug/mL while maintaining performance equivalent to regular fBS.
Catalog No. 50-290-4610 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F1283100ML
Animal serum is commonly used to supplement basal media formulations for the optimal growth of many cell types in vitro. fetal Bovine Serum (fBS) is the most common serum used to supplement cell culture media due to its high nutritional content. Although it is relatively low in protein fBS is effective in promoting and sustaining growth of vertebrate mammalian and insect cells. IgG is the most common immunoglobulin present in fBS. Elevated levels of IgG may cause problems within certain recombinant protein or monoclonal antibody purification processes. for applications where IgG may interfere or cause issues we have depleted IgG levels in this fetal bovine serum by sequential chromatography down to a level of 5 ug/mL while maintaining performance equivalent to regular fBS.