Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Fetal Bovine Serum, qualified, Canada

Catalog No. 12483020
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
Format:
Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
12-483-020 500 mL Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-483-020 Supplier Gibco™ Supplier No. 12483020

Introducing the new, aliquot-free Gibco™ One Shot™ FBS 50 mL bottle. Designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it. And that improves your chances for consistent results.

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category:Value (Standard)
  • Origin: Canada
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

Aliquot-free 50 mL bottle

The Gibco One Shot 50 mL bottle is designed to eliminate the need to aliquot, and the risk of contamination and variability that comes with it, improving your chances for consistent results.

Features include:

  • Wide mouth, chamfered edge, and curved shape for easy pouring and pipetting
  • Engineered to freeze/thaw without splitting or breaking
  • Fits in test tube racks and frees up freezer space
  • No labeling mishaps with our easy-to-read package label containing all of the information you need
  • 33% less waste generated than aliquoting from 500 mL bottles

All products may not be available in all regions due to importation regulations. Contact your local sales representative regarding product availability in your country.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Species Cattle/Bovine
Age Fetal
Country of Origin Canada
Format Bottle
Product Type Fetal Bovine Serum
Quantity 500 mL
Serum Treatment Standard (Sterile-filtered)
Sterility Sterile
Purity or Quality Grade Research Grade, Qualified
Shipping Condition Frozen
Show More Show Less

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.