Learn More
Omega Scientific Inc Fetal Bovine Serum, U.S. Source, 500 Ml
Catalog No. NC0570754
Encompass_Preferred
FB-11: Fetal Bovine Serum, U.S. SourceU.S. Source Fetal Bovine Sera is collected entirely from USDA licensed abattoirs and processed within the United States.-Virus Screened-Mycoplasma Tested-Triple 0.1 Micron Filtered-Low Hemoglobin lots (<15mg/dL) may be available.-Low Endotoxin lots (<1.0EU/mL) may be available.
Catalog No. NC0570754 Supplier Omega Scientific Inc Supplier No. FB11
FB-11: Fetal Bovine Serum, U.S. SourceU.S. Source Fetal Bovine Sera is collected entirely from USDA licensed abattoirs and processed within the United States.-Virus Screened-Mycoplasma Tested-Triple 0.1 Micron Filtered-Low Hemoglobin lots (<15mg/dL) may be available.-Low Endotoxin lots (<1.0EU/mL) may be available.