Learn More
Sigma Aldrich Fine Chemicals Biosciences fetal Bovine Serum USA ori
Catalog No. 502905104
Encompass
fBS remains a popular media supplement because it provides a wide array of functions in cell culture. fBS delivers nutrients growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SfM) systems.
Catalog No. 50-290-5104 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 12103C100ML
fBS remains a popular media supplement because it provides a wide array of functions in cell culture. fBS delivers nutrients growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SfM) systems.