Learn More
Sigma Aldrich Fine Chemicals Biosciences Fetal Bovine Serum USA ori1L
Catalog No. 502907316
Encompass
Fetal Bovine Serum (FBS) remains a popular media supplement as it provides a wide array of functions in cell culture. FBS delivers nutrients growth and attachment factors and aids in proliferation. It also protects cells from oxidative damage and apoptosis. FBS blocks reactive oxygen species generation which otherwise is not eliminated in serum-free media (SFM) systems.
Catalog No. 50-290-7316 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F41351L
Fetal Bovine Serum (FBS) remains a popular media supplement as it provides a wide array of functions in cell culture. FBS delivers nutrients growth and attachment factors and aids in proliferation. It also protects cells from oxidative damage and apoptosis. FBS blocks reactive oxygen species generation which otherwise is not eliminated in serum-free media (SFM) systems.