Learn More
Sigma Aldrich Fine Chemicals Biosciences fetal Bovine Serum USDA ap
Catalog No. 502900166
Encompass
fetal bovine serum (fBS) or fetal calf serum is the blood fraction remaining after the natural coagulation of blood. It comes from the blood drawn from a bovine fetus via a closed system of collection at the slaughterhouse. It is the most widely used serum-supplement for the in vitro cell culture of eukaryotic cells.
Catalog No. 50-290-0166 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F0926100ML
fetal bovine serum (fBS) or fetal calf serum is the blood fraction remaining after the natural coagulation of blood. It comes from the blood drawn from a bovine fetus via a closed system of collection at the slaughterhouse. It is the most widely used serum-supplement for the in vitro cell culture of eukaryotic cells.